Skip to Content
Merck
All Photos(1)

Documents

Z677000

ART® Softfit L Reload System

maximum volume 1000 μL, Tip design ART 1000L, Hinged rack, low retention: no

Synonym(s):

aerosol barrier tips, barrier filter pipette tip, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.01

material

colorless
polypropylene

sterility

sterile

feature

DNase free
RNase free
barrier
low retention: no
Hinged rack
Tip design ART 1000L

packaging

pack of 3072 ea (4 packs of 768 tips)

manufacturer/tradename

Molecular BioProducts 2779-HR

technique(s)

PCR: suitable

maximum volume

1000 μL

Looking for similar products? Visit Product Comparison Guide

General description

ART® Softfit L Reload System enables accurate and precise sample pipetting.
Designed to be compatible with LTS, the Molecular Bio Products Softfit L Reload System provides ART barrier pipette tips in color coded inserts by volume size. Choose from five pipette tip volume sizes with the option of low retention technology.

Each individual insert within the Softfit L Reload System is clearly identified with both product number and lot number allowing researchers to enjoy space-saving and environmental benefits of a reload tip system without loosing product traceability.

Features and Benefits

  • Hinged rack design enables convenient one-hand access to tips.
  • Color-coded inserts facilitate rapid identification of tip volume, simplifying the pipetting process.

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.
SoftFit-L is a trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service