Key Documents
Z673250
Stuart melting point tubes
Open both ends
About This Item
UNSPSC Code:
41121800
NACRES:
NB.79
Pricing and availability is not currently available.
material
soda-lime glass tube
sterility
non-sterile
feature
graduations
manufacturer/tradename
Stuart SMP1/4
diam.
1.3 mm
tube L
100 mm
tube O.D. × I.D. × wall thickness
1.9 mm × 1.3 mm × 0.3 mm
General description
Made from soda glass, these tubes are easy to seal in a Bunsen flame and break into two. Supplied in robust tube holders.
Legal Information
Stuart is a trademark of Bibby Sterlin Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)