Documents
Z655023
Carbolite® tube furnace end plugs
type D insulating plug, pack of 1
About This Item
UNSPSC Code:
41121800
NACRES:
NB.24
packaging
pack of 1
manufacturer/tradename
Carbolite EP-025-12B-001E
General description
For use on horizontal tube furnaces up to 1200°C only. Fits tubes up to 80 mm I.D.
For use with horizontal tube furnaces up to 1200C, with up to an 80mm ID process tube. Insulating plugs positioned at the ends of the process tube help prevent heat loss and improve uniformity in a tube furnace, a 6mm bore tube is fitted through the plug.
Legal Information
Carbolite is a registered trademark of Carbolite Limited