Documents
Z301108
Quickfit® expansion adapter
top female joint: ST/NS 24/29, bottom male joint: ST/NS 14/23
Synonym(s):
XA31
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Joint
female joint: ST/NS top24/29
male joint: ST/NS bottom14/23
manufacturer/tradename
Quickfit XA 31
availability
not available in USA and Canada
not available in USA and Canada
General description
These adapters connect items with larger cone to a piece of glassware with a smaller size socket.
Legal Information
Quickfit is a registered trademark of SciLabware Limited