Skip to Content
Merck
All Photos(1)

Documents

WHA13549204

Whatman® FF170HP Din A

sheets nitrocellulose membrane, pack of 10 sheets

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
NACRES:
NB.24

material

polyester backing
sheets nitrocellulose membrane

sterility

non-sterile

packaging

pack of 10 sheets

manufacturer/tradename

Whatman 13549204

size

210 m × 297 mm

thickness

200 μm

General description

FF High Performance (HP) membranes are part of the AE family, which are directly cast onto a plastic film. The FFHP membranes are a result of improved membrane casting procedures, which result in enhanced intra- and inter lot consistency and sharper lines. The surface is uniform without unincorporated nitrocellulose powder and the fine structure fiber distribution provides large internal surfaces for binding proteins

Nitrocellulose membranes are a key functional part of lateral-flow assays. The membrane must provide sufficient protein binding to enable the formation of a sharp and intense capture line, but at the same time the level of nonspecific background must be low for easy interpretation of results. Nitrocellulose membranes are available in a range of wicking rates and formulations. It has long been known that the wicking rate of a membrane has a significant impact on test sensitivity.

Features and Benefits

A backed and highly consistent membrane for use with low viscosity samples in a lateral flow assay.
  • Exceptional intra- and inter-lot consistency as demonstrated by a coefficient of variation (CV) for capillary rise of < 10%.</LI>Improved assay consistency and reproducible limit of detection.
  • Reduced assay optimization costs, compared to a less consistent membrane.

Legal Information

Whatman is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service