Skip to Content
Merck
All Photos(1)

Key Documents

W218502

Sigma-Aldrich

Isobutyl benzoate

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CO2CH2CH(CH3)2
CAS Number:
Molecular Weight:
178.23
FEMA Number:
2185
EC Number:
Council of Europe no.:
567
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.757

biological source

synthetic

grade

Kosher

reg. compliance

FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.495 (lit.)

bp

241-242 °C (lit.)

density

0.996 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

chocolate; rose

SMILES string

CC(C)COC(=O)c1ccccc1

InChI

1S/C11H14O2/c1-9(2)8-13-11(12)10-6-4-3-5-7-10/h3-7,9H,8H2,1-2H3

InChI key

KYZHGEFMXZOSJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Rationale for maximum allowable concentration of isobutyl benzoate in atmospheric air of populated areas].
M V Murkova et al.
Gigiena i sanitariia, (2)(2), 15-16 (1994-02-01)
Corine Tourné-Péteilh et al.
Physical chemistry chemical physics : PCCP, 13(34), 15523-15529 (2011-07-30)
Interfacial tension, electrical conductivity, NMR self-diffusion and DLS experiments have been used to investigate the self-aggregation in water of ionic liquids associating an ibuprofenate anion and 1-alkyl-3-methylimidazolium [C(n)MIm](+) (n = 4, 6, 8) cations. Despite the short alkyl chain on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service