Key Documents
TEM-HC400-AU-UL
Sigma-Aldrich
Ultra-Thin Holey Carbon Film Supported Gold Grids
size 400 mesh, box of 25
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Pricing and availability is not currently available.
packaging
box of 25
size
400 mesh
General description
A 4-6 nm ultra-thin holey carbon film supported grids. Gold grids, 400 Mesh, 25/box
Choose from one of the most recent versions:
Certificates of Analysis (COA)