Skip to Content
Merck
All Photos(2)

Documents

DWK241242008

Duran® Dropping Funnel

cylindrical funnel(s), graduated, standard ground cone 14/23, NS stopcock and key retaining device, capacity 50 mL

Synonym(s):

241242008

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121809
NACRES:
NB.43

material

cylindrical funnel(s)

feature

graduated

packaging

pkg of 1 ea

manufacturer/tradename

DURAN® 241242008

capacity

50 mL

Looking for similar products? Visit Product Comparison Guide

Application

uniform and metered liquid supply to a reaction mixture. The rate of supply can be adjusted.

Features and Benefits

With standard ground joint, standard ground stopcock and retaining device.

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service