CLS70320A50
Pyrex® VISTA™ round-bottom boiling flask
capacity 50 mL
Synonym(s):
boiling flask, bottom flask, round bottomed flask, Corning® 70320
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
round bottom flask
short
Joint
joint ST/NS19/38
sterility
non-sterile
feature
closure type Standard taper joint
packaging
pack of 2
case of 4
manufacturer/tradename
Corning 70320A-50
capacity
50 mL
neck size
(1)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
These round bottom boiling flasks have full length outer joints, but with shorter necks. Wall thicknesses are controlled to provide balance between thermal expansion and mechanical strength for greater shock resistance.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service