Documents
CLS49951L
Pyrex® wide-mouth graduated Erlenmeyer flask with screw-cap
capacity 1,000 mL
Synonym(s):
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer, Corning® 4995
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
polypropylene cap
screw top cap
wide-mouth
sterility
non-sterile
feature
cap
closure type Screw cap
packaging
case of 6
manufacturer/tradename
Corning 4995-1L
capacity
1,000 mL
General description
Features a GL 45 size, PP, plug-seal screw-cap on a wide, 29 mm neck opening that accommodates larger volume serological pipettes. Also good for media prep and storage, or as a culture flask when optional membrane cap is used.
Other Notes
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.