Skip to Content
Merck
All Photos(1)

Documents

796069

Sigma-Aldrich

13CO/H2/13CO2/N2 Gas Mixture

ratio (55:20:10:15), 99 atom % 13C, 99% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500

isotopic purity

99 atom % 13C

Assay

99% (CP)

form

gas

composition

ratio (55:20:10:15)

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Gas 1B - Press. Gas Compr. Gas - Repr. 1A - STOT RE 1 Inhalation

Target Organs

Heart

Storage Class Code

2A - Gases

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service