Skip to Content
Merck
All Photos(1)

Key Documents

680478

Sigma-Aldrich

Phenylboronic acid N-butyldiethanolamine ester

97%

Synonym(s):

6-Butyl-2-phenyl-1,3,6,2-dioxazaborocane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22BNO2
CAS Number:
Molecular Weight:
247.14
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

97%

form

solid

mp

126-130 °C

SMILES string

CCCCN1CCOB(OCC1)c2ccccc2

InChI

1S/C14H22BNO2/c1-2-3-9-16-10-12-17-15(18-13-11-16)14-7-5-4-6-8-14/h4-8H,2-3,9-13H2,1H3

InChI key

XBJLGTGASXIEFZ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service