Skip to Content
Merck
All Photos(1)

Documents

606286

Sigma-Aldrich

2-Phenyl-13C6-phenol

99 atom % 13C

Synonym(s):

[1,1′-Biphenyl]-2-ol-13C6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C6H10O
CAS Number:
Molecular Weight:
176.16
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

99 atom % 13C

bp

282 °C

mp

57-59 °C

mass shift

M+6

SMILES string

Oc1ccccc1-[13c]2[13cH][13cH][13cH][13cH][13cH]2

InChI

1S/C12H10O/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-9,13H/i1+1,2+1,3+1,6+1,7+1,10+1

InChI key

LLEMOWNGBBNAJR-QBTRQFLCSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

255.2 °F - closed cup

Flash Point(C)

124 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service