Skip to Content
Merck
All Photos(1)

Documents

600024

Sigma-Aldrich

2-Bromoethanol-13C2,1,1,2,2-d4

98 atom % D, 99 atom % 13C, 97% (CP)

Synonym(s):

Ethylene-13C2,d4 bromohydrin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br13CD213CD2OH
CAS Number:
Molecular Weight:
130.97
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C
98 atom % D

Assay

97% (CP)

technique(s)

protein expression: suitable

bp

56-57 °C/20 mmHg (lit.)

density

1.846 g/mL at 25 °C (lit.)

mass shift

M+5

SMILES string

Br[13C]([2H])([2H])[13C]([2H])([2H])O

InChI

1S/C2H5BrO/c3-1-2-4/h4H,1-2H2/i1+1D2,2+1D2

InChI key

LDLCZOVUSADOIV-URIFWVMJSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup - (own results)

Flash Point(C)

110 °C - closed cup - (own results)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service