Skip to Content
Merck
All Photos(1)

Key Documents

517038

Sigma-Aldrich

Titanium(IV) tetrahydrofurfuryloxide solution

50 wt. % in tetrahydrofurfuryl alcohol and isopropanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H36O8Ti
CAS Number:
Molecular Weight:
452.36
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

concentration

50 wt. % in tetrahydrofurfuryl alcohol and isopropanol

refractive index

n20/D 1.493

bp

178 °C

density

1.14 g/mL at 25 °C

SMILES string

C1COC(C1)CO[Ti](OCC2CCCO2)(OCC3CCCO3)OCC4CCCO4


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service