Skip to Content
Merck
All Photos(1)

Key Documents

410438

Sigma-Aldrich

Methyl (S)-(−)-3-Boc-2,2-dimethyl-4-oxazolidinecarboxylate

98%

Synonym(s):

Methyl (S)-(−)-3-(tert-butoxycarbonyl)-2,2-dimethyl-4-oxazolidinecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H21NO5
CAS Number:
Molecular Weight:
259.30
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

optical activity

[α]20/D −55°, c = 1.3 in chloroform

optical purity

ee: 99% (HPLC)

refractive index

n20/D 1.443 (lit.)

bp

101-102 °C/2 mmHg (lit.)

density

1.082 g/mL at 25 °C (lit.)

functional group

ester
ether

SMILES string

COC(=O)[C@@H]1COC(C)(C)N1C(=O)OC(C)(C)C

InChI

1S/C12H21NO5/c1-11(2,3)18-10(15)13-8(9(14)16-6)7-17-12(13,4)5/h8H,7H2,1-6H3/t8-/m0/s1

InChI key

ZNBUXTFASGDVCL-QMMMGPOBSA-N

Application

Precursor to a configurationally stable serinal derivative (Garner′s aldehyde) that has been used to make azasugars, sphingolipids, and the amino sugar found in calicheamicin, a potent antitumor agent.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kahne, D. et al.
Tetrahedron Letters, 31, 21-21 (1990)
Koskinen, A.M.P. et al.
Synlett, 665-665 (1990)
Garner, P. Park, J.M.
J. Org. Synth., 70, 18-18 (1992)
Dondoni, A. et al.
Journal of the Chemical Society. Chemical Communications, 1576-1576 (1991)
Garner, P. Park, J.M.
The Journal of Organic Chemistry, 52, 2361-2361 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service