Skip to Content
Merck
All Photos(1)

Key Documents

401005

Sigma-Aldrich

Cesium chromate

≥99.9% trace metals basis

Synonym(s):

Dicesium chromate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cs2CrO4
CAS Number:
Molecular Weight:
381.80
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

≥99.9% trace metals basis

form

powder

SMILES string

[Cs+].[Cs+].[O-][Cr]([O-])(=O)=O

InChI

1S/Cr.2Cs.4O/q;2*+1;;;2*-1

InChI key

BROHICCPQMHYFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Ox. Sol. 2 - Skin Sens. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service