Skip to Content
Merck
All Photos(1)

Key Documents

342106

Sigma-Aldrich

Diethyl azodicarboxylate

technical grade, 85%

Synonym(s):

DEAD

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCON=NCOOC2H5
CAS Number:
Molecular Weight:
174.15
Beilstein:
908662
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
Pricing and availability is not currently available.

grade

technical grade

Assay

85%

form

liquid

refractive index

n20/D 1.43 (lit.)

bp

106 °C/13 mmHg (lit.)

density

1.106 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)\N=N\C(=O)OCC

InChI

1S/C6H10N2O4/c1-3-11-5(9)7-8-6(10)12-4-2/h3-4H2,1-2H3/b8-7+

InChI key

FAMRKDQNMBBFBR-BQYQJAHWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exploding BombExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Expl. 1.1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

1 - Explosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jetze Visser et al.
Nature communications, 6, 6933-6933 (2015-04-29)
Despite intensive research, hydrogels currently available for tissue repair in the musculoskeletal system are unable to meet the mechanical, as well as the biological, requirements for successful outcomes. Here we reinforce soft hydrogels with highly organized, high-porosity microfibre networks that
Gregory S Watson et al.
Acta biomaterialia, 21, 109-122 (2015-03-17)
Geckos, and specifically their feet, have attracted significant attention in recent times with the focus centred around their remarkable adhesional properties. Little attention however has been dedicated to the other remaining regions of the lizard body. In this paper we
Adrian Egli et al.
The Journal of infectious diseases, 210(5), 717-727 (2014-03-13)
Feedback mechanisms between interferons α and λ (IFNs) may be affected by single nucleotide polymorphisms (SNP) in interleukin 28B (IL-28B; IFN-λ3) promoter region and may influence cytomegalovirus (CMV) replication. We associated IL-28B SNPs with the risk of CMV replication after
Fany Reffuveille et al.
Antimicrobial agents and chemotherapy, 58(9), 5363-5371 (2014-07-02)
Biofilm-related infections account for at least 65% of all human infections, but there are no available antimicrobials that specifically target biofilms. Their elimination by available treatments is inefficient since biofilm cells are between 10- and 1,000-fold more resistant to conventional
Victor Y L Leung et al.
Stem cells (Dayton, Ohio), 32(8), 2164-2177 (2014-04-17)
Intervertebral disc degeneration is associated with back pain and radiculopathy which, being a leading cause of disability, seriously affects the quality of life and presents a hefty burden to society. There is no effective intervention for the disease and the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service