Skip to Content
Merck
All Photos(1)

Key Documents

287849

Sigma-Aldrich

Tri(m-tolyl)phosphine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3C6H4)3P
CAS Number:
Molecular Weight:
304.37
Beilstein:
658863
EC Number:
MDL number:
UNSPSC Code:
12350000
PubChem Substance ID:

Assay

98%

mp

97-99 °C (lit.)

SMILES string

Cc1cccc(c1)P(c2cccc(C)c2)c3cccc(C)c3

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service