Skip to Content
Merck
All Photos(1)

Key Documents

275379

Sigma-Aldrich

2-Fluorothiophenol

97%

Synonym(s):

2-Fluorobenzenethiol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4SH
CAS Number:
Molecular Weight:
128.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

refractive index

n20/D 1.557 (lit.)

bp

61-62 °C/35 mmHg (lit.)

density

1.2 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Fc1ccccc1S

InChI

1S/C6H5FS/c7-5-3-1-2-4-6(5)8/h1-4,8H

InChI key

WJTZZPVVTSDNJJ-UHFFFAOYSA-N

Application

2-Fluorothiophenol was used in the synthesis of 2-bromo-2′-fluorodiphenyl sulfide[1].

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

118.0 °F - closed cup

Flash Point(C)

47.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of spiro (piperidine-4, 6'-dibenz [b, e]-1, 4-oxathiepin) and its 1-methyl derivative as potential antidepressant agents.
Collection of Czechoslovak Chemical Communications, 50(2), 503-509 (1985)
Homan Kang et al.
International journal of molecular sciences, 22(4) (2021-02-14)
We present a template-assisted method for synthesizing nanogap shell structures for biomolecular detections based on surface-enhanced Raman scattering. The interior nanogap-containing a silver shell structure, referred to as a silver nanogap shell (Ag NGS), was fabricated on silver nanoparticles (Ag
Homan Kang et al.
Scientific reports, 5, 10144-10144 (2015-05-29)
Recently, preparation and screening of compound libraries remain one of the most challenging tasks in drug discovery, biomarker detection, and biomolecular profiling processes. So far, several distinct encoding/decoding methods such as chemical encoding, graphical encoding, and optical encoding have been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service