Skip to Content
Merck
All Photos(1)

Key Documents

193674

Sigma-Aldrich

5-Nitro-1,10-phenanthroline

98%

Synonym(s):

Nitroferroin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H7N3O2
CAS Number:
Molecular Weight:
225.20
Beilstein:
196245
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

mp

202-204 °C (lit.)

SMILES string

[O-][N+](=O)c1cc2cccnc2c3ncccc13

InChI

1S/C12H7N3O2/c16-15(17)10-7-8-3-1-5-13-11(8)12-9(10)4-2-6-14-12/h1-7H

InChI key

PDDBTWXLNJNICS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service