Skip to Content
Merck
All Photos(1)

Key Documents

182451

Supelco

Polysulfone

average Mw 67,000 (Typical), average Mn 20,400 (Typical), pellets, secondary standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H4-4-C(CH3)2C6H4-4-OC6H4-4-SO2C6H4-4-O]n
CAS Number:
MDL number:
UNSPSC Code:
41116107

grade

secondary standard

form

pellets

autoignition temp.

1022 °F

mol wt

average Mn 20,400 (Typical)
average Mw 67,000 (Typical)

density

1.24 g/mL at 25 °C (lit.)

SMILES string

CC(C)(c1ccc(O)cc1)c2ccc(O)cc2.Oc3ccc(cc3)S(=O)(=O)c4ccc(O)cc4

Looking for similar products? Visit Product Comparison Guide

Application

Microwave cookware, appliance covers, surgical tools, automotive electrical components, printed circuit boards and plumbing components.

Features and Benefits

High impact resistance. Temperature use range up to 160°C. Exceptional steam sterilization stability. Electrical properties are maintained over a wide temperature and frequency range. High resistance to radiation degradation.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service