ADVH7F383B4E
AChemBlock
tert-butyl 3,3-difluoro-4-hydroxy-5-(hydroxymethyl)piperidine-1-carboxylate Cis-Trans mixture
About This Item
Iupac Name
tert-butyl 3,3-difluoro-4-hydroxy-5-(hydroxymethyl)piperidine-1-carboxylate
Inchi Code
1S/C11H19F2NO4/c1-10(2,3)18-9(17)14-4-7(5-15)8(16)11(12,13)6-14/h7-8,15-16H,4-6H2,1-3H3
InChI key
VPLQVCYHEZXTMT-UHFFFAOYSA-N
storage temp.
Purity
Country of Origin
Product Link
Synonym
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.achemblock.com/s89792-tert-butyl-3-3-difluoro-4-hydroxy-5-hydroxymethyl-piperidine-1-carboxylate-cis-trans-mixture.html
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service