- SmI2-promoted Reformatsky-type coupling reactions in exceptionally hindered contexts.
SmI2-promoted Reformatsky-type coupling reactions in exceptionally hindered contexts.
Organic letters (2008-02-28)
Brian A Sparling, Ryan M Moslin, Timothy F Jamison
PMID18302397
ABSTRACT
Highly substituted, very hindered enones were synthesized using a two-step procedure that utilizes a diiodosamarium-promoted Reformatsky-type coupling and dehydration using Martin sulfurane. Both alpha-chloro- and alpha-bromoketones were coupled with a variety of carbonyl nucleophiles to form the intermediate beta-hydroxyketones, occurring with excellent diastereoselectivity, favoring the syn isomer (R1=Me). This technique complements other methods and enables the preparation of enones outside of the scope of current olefination methodology.