Skip to Content
Merck
All Photos(1)

Key Documents

SAAR4164600

Methylene blue

redox indicator

Synonym(s):

Methylene Blue hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18ClN3S · xH2O
CAS Number:
Molecular Weight:
319.85 (anhydrous basis)
Beilstein:
3599847
MDL number:
UNSPSC Code:
41116100

Product Name

Methylene Blue,

packaging

pkg of 100 g (SAAR4164600DC)
poly bottle of 1 × 25 g (SAAR4164600CB)
poly bottle of 1 × 500 g (SAAR4164600EM)

storage temp.

2-30°C

SMILES string

O.[Cl-].CN(C)c1ccc2N=C3C=C\C(C=C3Sc2c1)=[N+](/C)C

InChI

1S/C16H18N3S.ClH.H2O/c1-18(2)11-5-7-13-15(9-11)20-16-10-12(19(3)4)6-8-14(16)17-13;;/h5-10H,1-4H3;1H;1H2/q+1;;/p-1

InChI key

WQVSELLRAGBDLX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

For laboratory use only

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service