SAAR190716
Hydrochloric acid
c(HCL) = 1.0 mol/l (1.0 N)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352106
Recommended Products
Product Name
Hydrochloric acid solution 0.1N (0.1M), density: 1.00 g/mL
packaging
poly bottle of 1 × 5 L (SAAR190716LL)
density
1.00 g/mL
storage temp.
2-30°C
Application
For laboratory use only
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible, corrosive hazardous materials
WGK
nwg
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service