Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.17
UNSPSC Code:
41121800
material
spherical glass
packaging
pkg of 1 L
manufacturer/tradename
Diversified Biotech BEAD-2100
technique(s)
PCR: suitable
size
4 mm (4.5 mm)
General description
Thermal-Micro Beads create a one-size-fits-all dry bath solution. Microbeads tightly surround tubes, microtubes and bottles to evenly distribute temperatures.
Features and Benefits
- Effectively heat and cool samples in most water and dry baths
- Reduce sample contamination in applications requiring water and ice
- Support tubes and bottles eliminating the need for floating racks and bottle weights
- Glass beads are more chemically stable and more easily cleanable than aluminum
- Thermal Microbeads are autoclavable and solvent resistant
- For use in water/dry baths, ice pans/buckets, Microbead Chambers and the Microbead Bag
Still not finding the right product?
Explore all of our products under Thermal Micro-Beads
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.