Z663085
Kern EW series Toploader balances
Kern EW 3000-2M + 963-127, DKD Calibration Certificate included
Synonym(s):
laboratory balance, weighing balance
About This Item
Recommended Products
AC/DC input
230 V
material
stainless steel weighing plate
feature
CE compliant
DKD Calibration Certificate included
packaging
pack of 1 ea
manufacturer/tradename
Kern EW 3000-2M + 963-127
availability
not available in USA and Canada
weighing capacity measuring range
3,000 g, resolution: 0.01 g, linearity: ±0.02 g
W × depth × H
180 mm × 230 mm × 85 mm
platform diam.
140 mm
weight
2 kg
battery power
rechargeable
Looking for similar products? Visit Product Comparison Guide
General description
- Large LCD display
- Capacity display
- Protective working cover
- Test weight
- 3 years warranty
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service