SYNA682440G
Synthware™ adapter for vacuum desiccator
with replacement for T-bore plug and retention kit plug 12/30mm
Synonym(s):
Synthware A682440G, glass flushing adapter
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
packaging
pack of 1 ea
manufacturer/tradename
Kemtech A682440G
General description
Replacement Adapter For Vacuum Desiccators.Includes Accessories: S510217 + S691230
Application
kemetch; synthware; vacuum adapter; desiccator
Features and Benefits
Organic Chemistry
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service