Skip to Content

SML3135

Cinobufagin

≥98% (HPLC)

Synonym(s):

14,15β-Epoxy-3β,16β-dihydroxy-5β,20(22)-bufadienolide 16-acetate, 5β,20(22)-Bufadienolide-3β,16β-diol-14,15β-epoxy 16-acetate, 14,15β-Epoxy-3β,16β-dihydroxy-5β,20(22)-bufadienolide 16-acetate, 5β,20(22)-Bufadienolide-3β,16β-diol-14,15β-epoxy 16-acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H34O6
CAS Number:
Molecular Weight:
442.54
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

C[C@]([C@@H](C(C=C1)=COC1=O)[C@H]2OC(C)=O)(CC[C@@]3([H])[C@@]4([H])CC[C@@]5([H])[C@@]3(CC[C@H](O)C5)C)[C@@]64[C@@H]2O6

InChI

1S/C26H34O6/c1-14(27)31-22-21(15-4-7-20(29)30-13-15)25(3)11-9-18-19(26(25)23(22)32-26)6-5-16-12-17(28)8-10-24(16,18)2/h4,7,13,16-19,21-23,28H,5-6,8-12H2,1-3H3/t16-,17+,18+,19-,21+,22-,23-,24+,25-,26-/m1/s1

InChI key

SCULJPGYOQQXTK-OLRINKBESA-N

Biochem/physiol Actions

Cinobufagin is a bufanolide steroid with anti-proliferative and pro-apoptotic effects as well as antinociception activity. It has been used in Chinese herbal medicine and has been shown to have clinical applications in a variety of cancers both as an anticancer agent and to reduce cancer pain. Cinobufagin was recently identified as novel expression-mimicker that exert in vitro as well as in vivo anti-AML efficacy against AML expressing mtRUNX1.


Still not finding the right product?

Explore all of our products under Cinobufagin


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library