Skip to Content
Merck

SML1777

Propacetamol hydrochloride

≥98% (HPLC), Paracetamol precursor, powder

Synonym(s):

4-Acetamidophenyl (diethylamino)acetate hydrochloride, Proparacetamol hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H20N2O3 · HCl
CAS Number:
Molecular Weight:
300.78
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Propacetamol hydrochloride, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

room temp

SMILES string

CC(NC1=CC=C(OC(CN(CC)CC)=O)C=C1)=O.[H]Cl

InChI

1S/C14H20N2O3.ClH/c1-4-16(5-2)10-14(18)19-13-8-6-12(7-9-13)15-11(3)17;/h6-9H,4-5,10H2,1-3H3,(H,15,17);1H

InChI key

WGTYJNGARJPYKG-UHFFFAOYSA-N

Biochem/physiol Actions

Propacetamol is a prodrug of paracetamol (acetaminophen) with improved water solubility.
Propacetamol is a prodrug of paracetamol (acetaminophen) with improved water solubility. Propacetamol is an analgesic drug delivered by IV when oral application of NSAID is not appropriate or contradicted.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library