Skip to Content
Merck

SML1492

UMI-77

≥98% (HPLC)

Synonym(s):

2-[[4-[[(4-Bromophenyl)sulfonyl]amino]-1-hydroxy-2-naphthalenyl]thio]-acetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H14BrNO5S2
CAS Number:
Molecular Weight:
468.34
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

OC1=C(SCC(O)=O)C=C(NS(C2=CC=C(Br)C=C2)(=O)=O)C3=CC=CC=C31

InChI

1S/C18H14BrNO5S2/c19-11-5-7-12(8-6-11)27(24,25)20-15-9-16(26-10-17(21)22)18(23)14-4-2-1-3-13(14)15/h1-9,20,23H,10H2,(H,21,22)

InChI key

WUGANDSUVKXMEC-UHFFFAOYSA-N

Biochem/physiol Actions

UMI-77 is an inhibitor of Induced myeloid leukemia cell differentiation protein Mcl-1, a member of the Bcl-2 family that inhibits apoptosis. UMI-77 acts by binding to the BH3-binding groove of Mcl-1, blocking the heterodimerization of Mcl-1/Bax and Mcl-1/Bak. UMI-77 is selective for Mcl-1 over other members of the antiapoptotic Bcl-2 family with a Ki of 490 nM. It inhibited cell growth and induced apoptosis in pancreatic cancer cells.
UMI-77 is an inhibitor of Induced myeloid leukemia cell differentiation protein Mcl-1.


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library