Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.84
UNSPSC Code:
12161503
Quality Segment
input
biological sample(s)
food(s)
plant sample(s)
application(s)
agriculture
cosmetics
environmental
food and beverages
detection method
colorimetric
relevant disease(s)
diabetes
storage temp.
−20°C
General description
Sorbitol, also known as glucitol, is a sugar alcohol metabolized slowly within the human body. It is derived from glucose through the reduction of an aldehyde group to a hydroxyl group. Excessive accumulation of sorbitol in erythrocytes, retinal cells, and Schwann cells has been linked to various health issues including retinopathy, cataracts, peripheral neuropathy, and diabetes. Sorbitol is primarily sourced from corn syrup and naturally occurs in fruits such as apples, pears, peaches, and prunes. Widely utilized as a sugar substitute and laxative, sorbitol also finds applications in specialty culture media, as well as in healthcare, food, and cosmetic products. Monitoring sorbitol levels in biological samples aids in understanding metabolic pathways and tracking the progression of diabetes.
Application
The Sorbitol Assay Kit may be used for:
- Metabolism Research
- Food and Beverage Analysis
- Diabetes Research
The kit is suitable for the quantitative determination of D-sorbitol and evaluation of drug effects on sorbitol metabolism in biological (e.g. blood), food, beverage and agriculture samples.
Biochem/physiol Actions
The sorbitol assay involves an end-point enzyme coupled MTT/NAD reaction that forms a colored product with an absorption maximum at A565 nm. The increase in absorbance at A565 nm is directly proportional to the sorbitol concentration.
Features and Benefits
- Fast and Precise Measurement: Achieve results in less than 1 hour, providing rapid analysis with a detection range of 5 - 1000 μM D-sorbitol ensures accurate measurements across a wide range of concentrations.
- High-Throughput Compatibility: Effortlessly integrate our kit into high-throughput handling systems, optimizing efficiency and ensuring accurate results.
Other Notes
For additional information on our range of Biochemicals, please complete this form.
Storage Class
12 - Non Combustible Liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.