Skip to Content
Merck
All Photos(1)

Key Documents

M303000

Sigma-Aldrich

CUTAG CPG

pore size 500 Å, 30-40 μmol/g

Synonym(s):

CUTAG CPG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.51

Quality Level

product line

Proligo Reagents

pore size

500 Å

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Legal Information

This product is covered by US Patent 7,202,264 owned by Isis Pharmaceuticals, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service