Merck
HS120791
Lab Drying Rack
white polystyrene (blue & green pegs)
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.12
UNSPSC Code:
41122800
Manufacturer/tradename:
Heathrow Scientific 120791
Feature:
to hold 36 pieces small glassware plus 36 pieces large glassware
Material:
white polystyrene (blue & green pegs)
Packaging:
pkg of 1 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
white polystyrene (blue & green pegs)
feature
to hold 36 pieces small glassware plus 36 pieces large glassware
packaging
pkg of 1 ea
manufacturer/tradename
Heathrow Scientific 120791
L × W × H
441 mm × 660 × 139 mm
General description
Included:-72 Pegs ∼ 36 Small (Green) ∼ 36 Large (Blue)-2 Drying Baskets-Drainage Tubing-Mounting Hardware
Features and Benefits
  • Most Versatile Rack in the market
  • Channels designed for more useful configurations
  • Large and Small pegs for all glassware varieties
  • One of a kind Drying Basket to collect smaller items
  • Designed to protect against breakage
  • Channels designed for more useful configurations
  • Peg angle allows for better hold - especially large, heavy pieces Patent Pending SureGrip Hook securely grips glassware into place
Choose from one of the most recent versions:
Certificates of Analysis (COA)