Skip to Content
Merck

H8041

IL-23 human

HumanKine®, recombinant, Xeno-free, >95% (SDS-PAGE), suitable for cell culture

Synonym(s):

Human IL-23, IL-23 Cytokine, Interleukin 23, Interleukin-23 human

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
Biological source:
human
Recombinant:
expressed in HEK 293 cells
Assay:
≥95% (SDS-PAGE)
Form:
lyophilized powder
Mol wt:
dimer 55 kDa (glycosylated)
Impurities:
≤1EU/mg
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Interleukin-23 human, IL-23, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture

biological source

human

Quality Segment

recombinant

expressed in HEK 293 cells

assay

≥95% (SDS-PAGE)

form

lyophilized powder

potency

≤2 ng/mL EC50

quality

endotoxin tested

mol wt

dimer 55 kDa (glycosylated)

packaging

pkg of 5X10 μg, pkg of 10 μg, pkg of 5 μg

technique(s)

cell culture | mammalian: suitable

impurities

≤1EU/mg

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL23A(51561)

General description

HumanKine interleukin-23 (IL-23) is expressed as a heterodimeric 55 kDa cytokine composed of two glycosylated and disulfide linked subunits (p40 cystine linked to p19). Production in human HEK 293 cells offers authentic glycosylation. Cytokines produced in Sf9 or Sf21 cells have post-translational modifications, which are not human-like.

Interleukin-23 (IL-23) is a pro-inflammatory cytokine. It is mostly synthesized by activated dendritic cells and macrophages. It is a 20.7kDa protein with 189 amino acids. IL-23 is mostly expressed on activated/memory T cells and natural killer (NK) cells. This gene is located on human chromosome 12q13.3.

Biochem/physiol Actions

Interleukin-23 (IL-23) plays a major role in cutaneous immune-mediated inflammatory diseases like psoriasis, allergic contact dermatitis and atopic dermatitis. It is involved in cellular immunity, driving the expansion, stabilization and survival of Th17 cells.

Analysis Note

The specific activity was determined by the dose-dependent secretion of h-IFN-γ or IL-8 from human PBMC.

Legal Information

HumanKine is a registered trademark of Proteintech Group, Inc. and Humanzyme, Inc


Still not finding the right product?

Explore all of our products under Interleukin-23 human


Storage Class

11 - Combustible Solids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library