Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41106514
Biological source:
human lung
Relevant disease(s):
cancer
Growth mode:
Adherent
Karyotype:
Modal no. 60
Morphology:
Epithelial
Manufacturer/tradename:
ECACC
biological source
human lung
manufacturer/tradename
ECACC
packaging
tube of 5 μg 95062823-DNA-5UG, pkg of vial of cells 95062823-1VL
growth mode
Adherent
karyotype
Modal no. 60
morphology
Epithelial
products
Variety of hormones, high level of calcitonin
receptors
Bombesin (BN), Epidermal growth factor (EGF), Transforming growth factor (TGF-beta), acetylcholine receptor (AChR)
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Application
Human lung tumour model
Biochem/physiol Actions
DMS 53 is a small cell carcinoma of the lung (SCLC) isolated in 1974 from a mediastinal biopsy at time of diagnosis from a 54 year-old male patient prior to treatment. DMS 53 secrete a large variety of hormones including adrenocorticotropin (ACTH), bombesin (BN), calcitonin (CT), chorionic gonadotrophin (hCG), glucagon, growth hormone (GH), oxytocin-neurophysin (OT-NP), parathyroid hormone (PTH), somatostatin-like immunoreactivity (SRIF), thyroid releasing hormone (TRH) and 17β-estradiol. Expression of cell surface antigens Leu-7, Class I HLA, CLASS II HLA and My23 (defined by AML-2-23 monoclonal antibody) has been shown. DMS 53 cells produce receptors for BN, epidermal growth factor (EGF), transforming growth factor-β (TGF β) and acetylcholine (AChR). This cell line was shown to be tumourigenic in nude, athymic mice.
Human Caucasian lung small cell carcinoma
STR-PCR Data: Amelogenin: X
CSF1PO: 12
D13S317: 10
D16S539: 12,13
D5S818: 10,11
D7S820: 8,11
THO1: 8,9.3
TPOX: 12
vWA: 15,17
CSF1PO: 12
D13S317: 10
D16S539: 12,13
D5S818: 10,11
D7S820: 8,11
THO1: 8,9.3
TPOX: 12
vWA: 15,17
Preparation Note
Split sub-confluent cultures (70-80%) 1:4 i.e. seeding at 4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Waymouth′s MB 752/1 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) (Heat Inactivated).
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). No licensing agreement is required when either this cell line or the DNA extracted from it are used for internal research purposes only. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to culturecollections@ukhsa.gov.uk.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.