Skip to Content
Merck

SYNPLE-BT004

Reagent-cartridge Biotin tag-PEG4-NH2

Synple

Synonym(s):

Synple 2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12161503
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

reaction suitability

reaction type: Biotinylations

parameter

up to 0.1 mmol scale

SMILES string

[s]1cnc(c1c2ccc(cc2)CNC(=O)[C@H]3N(C[C@@H](C3)O)C(=O)[C@@H](NC(=O)CCOCCC(=O)O)C(C)(C)C)C

InChI

1S/C28H38N4O7S/c1-17-24(40-16-30-17)19-7-5-18(6-8-19)14-29-26(37)21-13-20(33)15-32(21)27(38)25(28(2,3)4)31-22(34)9-11-39-12-10-23(35)36/h5-8,16,20-21,25,33H,9-15H2,1-4H3,(H,29,37)(H,31,34)(H,35,36)/t20-,21+,25-/m1/s1

InChI key

HVPZHRHEHKZOOJ-TYBLODHISA-N

General description

Reagent-cartridge Biotin tag-PEG4-NH2 is a reagent cartridge for the Synple 2 Automated Synthesizer that enables the biotin tagging of aldehydes and ketones through a reductive amination. The cartridge contains all necessary reagents for linking the chosen aldehyde or ketone substrate to biotin.

Scale: up to 0.1 mmol
Linker: PEG4
Terminal functional group: amine
Compatible substrates: aldehyde, ketone

Cartridge contents:
  • Reducing agent (supported cyanoborohydride)
  • Base (supported triethylamine)
  • SCX (for purification)
  • Biotin linker

Related Cartridges:

  • SYNPLE-BT002Reagent-cartridge Biotin tag-PEG2-NH2
  • SYNPLE-BT003Reagent-cartridge Biotin tag-PEG3-NH2
  • SYNPLE-BT012Reagent-cartridge Biotin tag-PEG2-O-4-P-NH
  • SYNPLE-BT013Reagent-cartridge Biotin tag-PEG3-O-4-P-NH
  • SYNPLE-BT014Reagent-cartridge Biotin tag-PEG4-O-4-P-NH
  • SYNPLE-BT022Reagent-cartridge Biotin tag-PEG2-a-COOTfp
  • SYNPLE-BT023Reagent-cartridge Biotin tag-PEG3-a-COOTfp
  • SYNPLE-BT024Reagent-cartridge Biotin tag-PEG4-a-COOTfp
  • SYNPLE-BT032Reagent-cartridge Biotin tag-PEG2-COOTfp
  • SYNPLE-BT033Reagent-cartridge Biotin tag-PEG3-COOTfp
  • SYNPLE-BT034Reagent-cartridge Biotin tag-PEG4-COOTfp

Learn more about the Synple automated synthesis platform.

Features and Benefits of Synple Platform
  • Time savings - pre-installed protocols have been highly optimized, which obviates the need for the user′s own reaction development and optimization
  • Easy to use - protocols are loaded by simply scanning the cartridge; does not requires any programming knowledge
  • Safety - all chemicals are contained in the system, minimizing the user′s exposure
  • Set-up reaction in less than 5 minutes
  • Small footprint
  • Plug & Play system - simple installation
  • Reaction temperatures from 25 °C - 80 °C
  • Cartridges available for many different reactions
  • Prefilled with all required reagents
  • RFID chip containing optimized reaction protocol

Legal Information

Product of Synple Chem AG


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library