Skip to Content
Merck

PHL89752

Patchouli alcohol

phyproof® Reference Substance

Synonym(s):

(1R,3R,6S)-2,2,6,8-Tetramethyltricyclo[5.3.1.03,8]undec-3-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H26O
CAS Number:
Molecular Weight:
222.37
NACRES:
NA.24
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
2553847
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

primary reference standard

product line

phyproof® Reference Substance

assay

≥98.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

CC1(C)[C@@H]2CC[C@]3(C)[C@@]1(O)CC[C@H](C)[C@@H]3C2

InChI

1S/C15H26O/c1-10-5-8-15(16)13(2,3)11-6-7-14(15,4)12(10)9-11/h10-12,16H,5-9H2,1-4H3/t10-,11?,12?,14-,15?/m0/s1

InChI key

GGHMUJBZYLPWFD-DUNKBJDJSA-N

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Other Notes

This compound is commonly found in plants of the genus: valeriana

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG


Still not finding the right product?

Explore all of our products under Patchouli alcohol


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library