Skip to Content
Merck

K2000010

Ketoprofen impurity A

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-(3-Benzoylphenyl)ethanone, 3-Acetylbenzophenone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H12O2
CAS Number:
Molecular Weight:
224.25
UNSPSC Code:
41116107
NACRES:
NA.24
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

ketoprofen

manufacturer/tradename

EDQM

application(s)

pharmaceutical
pharmaceutical (small molecule)

format

neat

SMILES string

O=C(C)c1cc(ccc1)C(=O)c2ccccc2

InChI

1S/C15H12O2/c1-11(16)13-8-5-9-14(10-13)15(17)12-6-3-2-4-7-12/h2-10H,1H3

InChI key

PPNGALBGZJBTRY-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ketoprofen impurity A EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.


Pictograms

Environment

Hazard Codes

Hazard Classifications

Aquatic Chronic 2

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library