Skip to Content

38595

α-(4-Chlorophenyl)-α-[2-(dimethylamino)ethyl]-2-pyridineacetonitrile hydrochloride

pharmaceutical impurity standard

Synonym(s):

Chlorpheniramine impurity D (PhEur), (2RS)-2-(4-Chlorophenyl)-4-dimethylamino-2-(2-pyridyl)butyronitril hydrochloride, Chlorpheniramine nitrile hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H18ClN3 · HCl
Molecular Weight:
336.26
UNSPSC Code:
41116107
NACRES:
NA.24
Beilstein/REAXYS Number:
268419
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

API family

chlorpheniramine

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤1.0% water

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

ClC(C=C1)=CC=C1C(CCN(C)C)(C#N)C2=NC=CC=C2.Cl



pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library