Skip to Content
Merck

09846

Boc-β-Ala-PAM resin

extent of labeling: ~0.5 mmol/g loading

Synonym(s):

Boc-β-alanine bound to PAM resin, PAM resin esterified with Boc-β-alanine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

beads

Quality Segment

reaction suitability

reaction type: Boc solid-phase peptide synthesis

extent of labeling

~0.5 mmol/g loading

matrix

crosslinked with 1% DVB

particle size

100-200 mesh

application(s)

peptide synthesis

Other Notes

Resin for the synthesis of Multiple Antigenic Peptides, MAP


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library