Skip to Content
Merck

Skip To

MDI02ADAP

Milli-DI® 0.2 μm Filter Adapter

The adapter allows to connect a Millipak® final filter

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41104214
NACRES:
JA.14
eCl@ss:
33050190
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 1 unit

manufacturer/tradename

Milli-DI®

compatibility

for use with Milli-DI®

shipped in

ambient

General description

Milli-DI® 0.2 μm filter adapter is a water purification accessory for use with Milli-DI® Water Purification System. It is connected to the tubing, and then to the water supply of the system.

Legal Information

MILLI-DI is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIPAK is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
0PTIA04NN1CF0F012P5TANKV01A1
compatibility

for use with Milli-DI®

compatibility

for use with Milli-Q® CLX

compatibility

for use with AFS® (1999-2012)

compatibility

for use with Milli-Q® EQ 7008, for use with Milli-Q® EQ 7016, for use with Milli-Q® IX 7003, for use with Milli-Q® IX 7005, for use with Milli-Q® IX 7010, for use with Milli-Q® IX 7015, for use with Milli-Q® IQ 7003, for use with Milli-Q® IQ 7005, for use with Milli-Q® IQ 7010, for use with Milli-Q® IQ 7015

shipped in

ambient

shipped in

-

shipped in

ambient

shipped in

ambient

packaging

pkg of 1 unit

packaging

pkg of 1 unit

packaging

pkg of 1 kit

packaging

pkg of 1 unit

manufacturer/tradename

Milli-DI®

manufacturer/tradename

Opticap®

manufacturer/tradename

Milli-Q®

manufacturer/tradename

-




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters