Skip to Content
Merck

MABC1823

Anti-CD271/p75NTR Antibody, clone G4B1

Synonym(s):

CD271, Gp80-LNGFR, Low affinity neurotrophin receptor p75NTR, Low-affinity nerve growth factor receptor, Tumor necrosis factor receptor superfamily member 16, p75 ICD

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.46
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
G4B1, monoclonal
Application:
ELISA, FACS, ICC, IHC, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

G4B1, monoclonal

mol wt

calculated mol wt 45.18 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100

technique(s)

ELISA: suitable, flow cytometry: suitable, immunocytochemistry: suitable, immunohistochemistry: suitable, western blot: suitable

isotype

IgG2aκ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... NGFR(4804)

General description

Tumor necrosis factor receptor superfamily member 16 (UniProt: P08138; also known as Gp80-LNGFR, Low affinity neurotrophin receptor p75NTR, Low-affinity nerve growth factor receptor, p75 ICD, CD271) is encoded by the NGFR (also known as TNFRSF16) gene (Gene ID: 4804) in human. CD271 is single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-28), which is subsequently cleaved off to produce the mature form that contains an extracellular domain (aa 29-250), a transmembrane domain (aa 251-272), and a cytoplasmic domain (aa 273-427). It serves as a low affinity receptor that can bind to NGF, BDNF, NTF3, and NTF4. It forms a heterodimeric receptor with sortilin related VPS10 domain containing receptor 2 (SorCS2) that binds the precursor forms of NGF, BDNF and NTF3 with high affinity, but has much lower affinity for mature NGF and BDNF. CD271 plays an important role in differentiation and survival of specific neuronal populations during development. It can mediate cell survival as well as cell death of neural cells. During development, it signals toward apoptosis to eliminate unnecessary neurons. Its death domain is localized to amino acids 344-421, which is responsible for its interaction with RAN-binding protein 9 (RANBP9). Neurotrophin binding to p75 can activate the MAP kinase pathway leading to cell survival or apoptosis depending on the cell type or ligand type. Clone G4B1 is shown to bind to the fourth cysteine-rich domain (CRD4) in the extracellular domain and is specifically detects glycosylated CD271. It recognizes CD271 expressed in esophageal cancer, but not in normal esophageal basal cells. It also detects the fetal esophageal epithelium and Barrett′s esophagus that possess stem cell-like characteristics. (Ref.: Fujii, K., et al. (2022). Cancer Sci. 113(8); 2878-2887; Glerup, S., et al. (2014). Neuron. 82(5); 1074-1087; Baeza-Raja , B., et al. (2013). J Neurosci. 33(25); 10221-10234).

Immunogen

A recombinant fragment corresponding to the extracellular domain of human CD271/p75NTR.

Application

Quality Control Testing

Evaluated by Flow Cytometry in HPCM2 cells.

Flow Cytometry Analysis: 1.0 µg of this antibody detected CD271/p75NTR in one million HPCM2 cells.

Tested Applications

Immunocytochemistry Analysis: A representative lot detected CD271 in Immunocytochemistry applications (Fujii, K., et al. (2022). Cancer Sci. 113(8):2878-2887).

Flow Cytometry Analysis: A representative lot detected CD271 in Flow Cytometry applications (Fujii, K., et al. (2022). Cancer Sci. 113(8):2878-2887).

Immunohistochemistry Applications: A representative lot detected CD271/p75NTR in Immunohistochemistry applications (Fujii, K., et al. (2022). Cancer Sci. 113(8):2878-2887).

Western Blotting Analysis: A representative lot detected CD271 in Western Blotting applications (Fujii, K., et al. (2022). Cancer Sci. 113(8):2878-2887).

ELISA Analysis: A representative lot detected CD271 in ELISA applications (Fujii, K., et al. (2022). Cancer Sci. 113(8):2878-2887).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone G4B1 is a mouse monoclonal antibody that detects CD271 (p75ICD). It targets an epitope within the fourth cysteine-rich domain in the extracellular domain.

Physical form

Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library