Skip to Content
Merck

AG240

Synuclein α, control peptide for AB5334P

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.28
eCl@ss:
32160702
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

form

solid

manufacturer/tradename

Chemicon®

technique(s)

cell based assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... SNCA(6622)

General description

Peptide corresponding to sequence numbers 116-131of the human alpha synuclein molecule.

Application

For use in blocking the reactivity of AB5334P.

Optimal working dilution must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Physical form

Lyophilized. Reconstitute with 100 μL of sterile distilled water. Centrifuge to remove insoluble material. Contains no preservative.

Preparation Note

Maintain lyophilized material at 2-8°C for up to 6 months. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library