Skip to Content

8.40068

Tetraoctylphosphonium bromide

for synthesis

Synonym(s):

Tetraoctylphosphonium bromide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C32H68BrP
CAS Number:
Molecular Weight:
563.76
MDL number:
UNSPSC Code:
12352107
EC Index Number:
245-935-7
NACRES:
NA.22
Assay:
≥95.0%
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95.0%

form

solid

potency

2800 mg/kg LD50, oral (Rat)

reaction suitability

core: phosphonium

pH

<3 (50 g/L in H2O)

mp

40-45 °C

transition temp

flash point 121 °C

storage temp.

2-30°C

SMILES string

[P+](CCCCCCCC)(CCCCCCCC)(CCCCCCCC)CCCCCCCC.[Br-]

InChI

1S/C32H68P.BrH/c1-5-9-13-17-21-25-29-33(30-26-22-18-14-10-6-2,31-27-23-19-15-11-7-3)32-28-24-20-16-12-8-4;/h5-32H2,1-4H3;1H/q+1;/p-1

InChI key

QVBRLOSUBRKEJW-UHFFFAOYSA-M

Analysis Note

Assay (argentometric): ≥ 95.0 %
Identity (IR): passes test


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 3

flash_point_f

249.8 °F - closed cup

flash_point_c

121 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library