Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
eCl@ss:
32011401
UNSPSC Code:
12352202
NACRES:
NA.25
Biological source:
bovine serum
Mol wt:
mean molar mass 188.34 g/mol
Quality Level
biological source
bovine serum
grade
synthesis grade
type
for cell culture
potency
175 mg/kg LD50, oral, 350 mg/kg LD50, oral
shelf life
36 mo.
mol wt
mean molar mass 188.34 g/mol
description
(nitrile rubber), 191 °C (flame point), >200 °C (ignition temperature)
pH
6 (20 °C, 0.2 g/L in H2O)
bp
>250 °C (1013 hPa)
mp
25-27 °C
density
1.05 g/cm3 at 20 °C
storage temp.
2-8°C
General description
Purified from bovine serum through the use of serum acidification and organic solvents. Treated to remove proteases. This product will have a molecular mass of 150 – 200kDa. BGG is offered as a low salt, lyophilized, off-white powder.
Preparation Note
refrigerated
Disclaimer
After skin contact: wash off with plenty of water. Remove contaminated clothing.
For further manufacturing use only. Not for human or animal consumption.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.