Skip to Content
Merck

8.06766

4-Nitrobenzaldehyde

for synthesis

Synonym(s):

4-Nitrobenzaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
4-(NO2)C6H4CHO
CAS Number:
Molecular Weight:
151.12
UNSPSC Code:
12352114
EC Index Number:
209-084-5
NACRES:
NA.22
MDL number:
Assay:
≥98.0% (GC)
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98.0% (GC)

form

solid

potency

4700 mg/kg LD50, oral (Rat), 16000 mg/kg LD50, skin (Rat)

mp

101-104 °C

bulk density

400 kg/m3

storage temp.

2-30°C

SMILES string

[N+](=O)([O-])c1ccc(cc1)C=O

InChI

1S/C7H5NO3/c9-5-6-1-3-7(4-2-6)8(10)11/h1-5H

InChI key

BXRFQSNOROATLV-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 103 °C
Melting range (upper value): ≤ 108 °C
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under 4-Nitrobenzaldehyde


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library