Skip to Content
Merck

71713

D-Tube96 Dialyzer, 12-14 kDa

Synonym(s):

Dialysis Tubing

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.24
UNSPSC Code:
41104900
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

Novagen®

Quality Segment

storage condition

OK to freeze

shipped in

ambient

storage temp.

10-30°C

General description

The D-Tube96 Dialyzer allows convenient and simultaneous dialysis of 96 samples in a high throughput manner. The device takes advantage of the features of the D-Tube Dialyzer Mini in a 96-tube configuration. D-Tube Dialyzers are easy to handle dialyzers with dual membranes and large surface area. The membrane is ultra clean, EDTA-treated regenerated cellulose, sulfur- and heavy metal- free and available in molecular weight cutoffs of 6-8 kDa or 12-14 kDa. The D-Tube Dialyzers Mini have a capacity of 10-250 µl and are ideal for small samples. The samples are added and removed using a standard laboratory pipet, multichannel pipet, or by robotic dispensing. D-Tube96 Dialyzers are suitable for dialysis of proteins, oligonucleotide, RNA, and DNA in a high-throughput format. Each kit contains a floating rack with 96 D-Tube Dialyzers Mini, 96 caps and an adhesive aluminum plate sealer. If fewer than 96 tubes are required, excess tubes can be removed from the modular device and saved for use at a later time. D-Tube96 Dialyzers are ideally suited for buffer exchange of protein samples into a physiologically relevant buffer after screening for optimal refolding conditions with the iFOLD Protein Refolding Systems.

Other Notes

•: 1: D-Tube96 Dialyzer, 12-14 kDa


•: 96: D-Tube96 Dialyzer caps


•: 1: Aluminum Plate Sealer

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

10-13 - German Storage Class 10 to 13



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library