5.09106
Sphingosine Kinase-2 Inhibitor, K145
Synonym(s):
Sphingosine Kinase-2 Inhibitor, K145, SphK2 Inhibitor, K145
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C18H24N2O3S
CAS Number:
Molecular Weight:
348.46
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Recommended Products
Assay
≥95% (HPLC)
Quality Level
form
powder
potency
4.3 μM IC50
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
off-white
solubility
DMSO: 100 mg/mL
water: 2.5 mg/mL
storage temp.
2-8°C
General description
A cell permeable, orally bioavailable thiazolidine-2,4-dione derivative that acts as a selective, sphingosine competitive inhibitor of spingosine Kinase -2 (SphK2; IC50 = 4.3 µM; Ki = 6.4 µM), but does not affect the activities of sphingosine kinase-1, ceremide kinase, and ceramide synthase even at high concentration (~10 micro;M). Shown to inhibit the growth of U937 human leukemic monocyte lymphoma cells by inducing apoptosis and inhibiting the phosphorylation of ERK and Akt. Blocks the growth of U937 cells in nude mice and inhibits the growth of JC tumor cells in BALB/c mice without any apparent toxicity.
Please note that the molecular weight for this compound is batch-specific due to variable water content.
Please note that the molecular weight for this compound is batch-specific due to variable water content.
Biochem/physiol Actions
Cell permeable: yes
Primary Target
Sphk2
Sphk2
Packaging
Packaged under inert gas
Warning
Toxicity: Standard Handling (A)
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 months at -20°C.
Other Notes
Liu, K., et al. 2013. PLos One8, e56471.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service